Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007346.2 | NP_031372.2 | ||||
GenBank mRNA | AF109134.1 | - | ||||
AF172449.1 | - | |||||
AF172450.1 | - | |||||
AF172451.1 | - | |||||
AF172452.1 | - | |||||
AF172453.1 | - | |||||
AK022234.1 | - | |||||
AK024485.1 | - | |||||
AK314696.1 | - | |||||
BC008768.2 | - | 3050950 | ||||
BC014137.2 | - | 4643904 | ||||
BC032666.1 | - | 5552531 | ||||
BM551600.1 | - | |||||
ENST00000290291.0 | - |