Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001278601.1 | NP_001265530.1 | ||||
NM_013693.3 | NP_038721.1 | |||||
GenBank mRNA | AK153319.1 | - | ||||
AK153800.1 | - | |||||
AK154223.1 | - | |||||
AK155964.1 | - | |||||
AY423855.1 | - | |||||
BC117057.1 | - | 40126376 | ||||
BC137720.1 | - | 8860739 | ||||
BY167004.1 | - | |||||
M11731.1 | - | |||||
M13049.1 | - | |||||
X02611.1 | - |