Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
GenBank mRNA | AF128210.1 | - | ||||
AF128211.1 | - | |||||
AF128212.1 | - | |||||
AF128213.1 | - | |||||
AK153729.1 | - | |||||
AV363060.1 | - | |||||
BC116851.1 | - | |||||
BC116855.1 | - | |||||
M86672.1 | - |