Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001111099.1 | NP_001104569.1 | ||||
NM_007669.4 | NP_031695.1 | |||||
GenBank mRNA | AB017817.1 | - | ||||
AB017818.1 | - | |||||
AK007630.1 | - | |||||
AK089093.1 | - | |||||
AK139306.1 | - | |||||
AK149910.1 | - | |||||
AK170467.1 | - | |||||
AK194916.1 | - | |||||
AK202927.1 | - | |||||
AK210703.1 | - | |||||
BC002043.1 | - | 3495942 | ||||
BY206886.1 | - | |||||
CT010218.1 | - | |||||
U09507.1 | - | |||||
U24173.1 | - |