Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001111274.1 | NP_001104744.1 | ||||
NM_001111275.1 | NP_001104745.1 | |||||
NM_001111276.1 | NP_001104746.1 | |||||
NM_010512.4 | NP_034642.2 | |||||
NM_184052.3 | NP_908941.1 | |||||
GenBank mRNA | AF440694.1 | - | ||||
AK038119.1 | - | |||||
AK050118.1 | - | |||||
AK052033.1 | - | |||||
AK081019.1 | - | |||||
AK155435.1 | - | |||||
AK165471.1 | - | |||||
AY878192.1 | - | |||||
AY878193.1 | - | |||||
BC012409.1 | - | 4194295 | ||||
CA494190.1 | - | |||||
CT010364.1 | - | |||||
X04480.1 | - | |||||
X04482.1 | - |