Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007527.3 | NP_031553.1 | ||||
GenBank mRNA | AK013298.1 | - | ||||
AK149994.1 | - | |||||
AK159233.1 | - | |||||
AV355155.1 | - | |||||
AY095934.1 | - | |||||
BC018228.1 | - | 3968903 | ||||
BC053380.1 | - | 6489475 | ||||
BY308683.1 | - | |||||
CJ078904.1 | - | |||||
L22472.1 | - |