Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001136067.2 | NP_001129539.1 | ||||
NM_001161421.1 | NP_001154893.1 | |||||
NM_001161422.1 | NP_001154894.1 | |||||
NM_001161423.1 | NP_001154895.1 | |||||
NM_001161424.1 | NP_001154896.1 | |||||
NM_010547.2 | NP_034677.2 | |||||
NM_178590.4 | NP_848705.1 | |||||
GenBank mRNA | AF069542.1 | - | ||||
AF513109.1 | - | |||||
AK037020.1 | - | |||||
AK042138.1 | - | |||||
AK047400.1 | - | |||||
AK048446.1 | - | |||||
AK148163.1 | - | |||||
AK154095.1 | - | |||||
AK188269.1 | - | |||||
AY112937.1 | - | |||||
BC021431.1 | - | 5030381 | ||||
BC026905.1 | - | |||||
BY768708.1 | - | |||||
CJ099503.1 | - | |||||
CN701499.1 | - | |||||
CN717929.1 | - | |||||
CV556906.1 | - |