Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001163530.1 | NP_001157002.1 | ||||
NM_011164.2 | NP_035294.2 | |||||
GenBank mRNA | AK017520.1 | - | ||||
AK017521.1 | - | |||||
AK017547.1 | - | |||||
AK017562.1 | - | |||||
AK017579.1 | - | |||||
AK017584.1 | - | |||||
AK077493.1 | - | |||||
AK133645.1 | - | |||||
AK133736.1 | - | |||||
AK133752.1 | - | |||||
AK161553.1 | - | |||||
AK161557.1 | - | |||||
AK161565.1 | - | |||||
AK161594.1 | - | |||||
AK161619.1 | - | |||||
BC061141.1 | - | 30250937 | ||||
X02892.1 | - | |||||
X04418.1 | - |