Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001163815.1 | NP_001157287.1 | ||||
NM_001163816.1 | NP_001157288.1 | |||||
NM_011691.4 | NP_035821.3 | |||||
GenBank mRNA | AK088586.1 | - | ||||
AK132886.1 | - | |||||
AK146347.1 | - | |||||
AK150398.1 | - | |||||
AK151183.1 | - | |||||
AK151214.1 | - | |||||
AK151367.1 | - | |||||
AK151373.1 | - | |||||
AK151831.1 | - | |||||
AK152073.1 | - | |||||
AK152425.1 | - | |||||
AK152555.1 | - | |||||
AK153052.1 | - | |||||
AK153265.1 | - | |||||
AK154183.1 | - | |||||
AK159326.1 | - | |||||
AK169478.1 | - | |||||
AK171873.1 | - | |||||
BC020487.1 | - | 3965451 | ||||
BY139350.1 | - | |||||
BY496946.1 | - | |||||
CT010321.1 | - | |||||
D83266.1 | - | |||||
M59833.1 | - | |||||
X64361.1 | - |