Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_011785.3 | NP_035915.3 | ||||
GenBank mRNA | AF124142.1 | - | ||||
AK052953.1 | - | |||||
AK148914.1 | - | |||||
BC066861.1 | - | 30089997 | ||||
CK390299.1 | - |