Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001081279.1 | NP_001074748.1 | ||||
GenBank mRNA | AK010068.1 | - | ||||
AK035252.1 | - | |||||
AK081646.1 | - | |||||
AK132349.1 | - | |||||
AK171731.1 | - | |||||
AK171759.1 | - | |||||
BC082308.1 | - |