Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_019777.3 | NP_062751.2 | ||||
GenBank mRNA | AB016589.1 | - | ||||
AK088580.1 | - | |||||
AK149911.1 | - | |||||
AK153046.1 | - | |||||
AK180824.1 | - | |||||
AK193494.1 | - | |||||
AK198423.1 | - | |||||
AK209824.1 | - | |||||
AK213678.1 | - | |||||
AK218764.1 | - | |||||
AK220324.1 | - | |||||
BC037446.1 | - | 3980145 | ||||
CT010206.1 | - | |||||
CT010313.1 | - |