Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_174885.3 | NP_777479.2 | ||||
GenBank mRNA | AY158689.1 | - | ||||
BC048757.1 | - | |||||
BC108387.1 | - | |||||
BC139059.1 | - | 8862081 | ||||
KF144375.1 | - | |||||
KF144376.1 | - |