Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007912.4 | NP_031938.1 | ||||
NM_207655.2 | NP_997538.1 | |||||
GenBank mRNA | AF124513.1 | - | ||||
AF275367.1 | - | |||||
AF277898.1 | - | |||||
AK004883.1 | - | |||||
AK004911.1 | - | |||||
AK004944.1 | - | |||||
AK014017.1 | - | |||||
AK033431.1 | - | |||||
AK049452.1 | - | |||||
AK087861.1 | - | |||||
AK134262.1 | - | |||||
AK163338.1 | - | |||||
BC023729.1 | - | 5351807 | ||||
BC030681.1 | - | |||||
BQ553815.1 | - | |||||
BY725722.1 | - | |||||
L06864.1 | - | |||||
U03425.1 | - | |||||
X59698.1 | - | |||||
X78987.1 | - | |||||
Z12608.1 | - |