Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008008.4 | NP_032034.1 | ||||
GenBank mRNA | AK033934.1 | - | ||||
AK037172.1 | - | |||||
AK086639.1 | - | |||||
AK139776.1 | - | |||||
BC052847.1 | - | 30054875 | ||||
Z22703.1 | - |