Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_009506.2 | NP_033532.1 | ||||
GenBank mRNA | AK047844.1 | - | ||||
AK216473.1 | - | |||||
BC096377.1 | - | 5002720 | ||||
EU862553.1 | - | |||||
EU862554.1 | - | |||||
EU862555.1 | - | |||||
U58112.1 | - | |||||
U73620.1 | - |