Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008542.3 | NP_032568.3 | ||||
GenBank mRNA | AB221689.1 | - | ||||
AF010133.1 | - | |||||
AI415735.1 | - | |||||
AK004671.1 | - | |||||
AK046602.1 | - | |||||
BC047280.1 | - | 6477199 |