Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007553.3 | NP_031579.2 | ||||
GenBank mRNA | AK132433.1 | - | ||||
AK133923.1 | - | |||||
AK142118.1 | - | |||||
AK161862.1 | - | |||||
AV333272.1 | - | |||||
BC100344.1 | - | 30846365 |