Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008928.4 | NP_032954.1 | ||||
GenBank mRNA | AK008141.1 | - | ||||
AK011002.1 | - | |||||
AK053378.1 | - | |||||
AK150067.1 | - | |||||
AK159379.1 | - | |||||
AK166255.1 | - | |||||
AK166507.1 | - | |||||
AK167209.1 | - | |||||
AK167442.1 | - | |||||
AK168710.1 | - | |||||
AK176968.1 | - | |||||
AK179609.1 | - | |||||
AK181928.1 | - | |||||
AK185191.1 | - | |||||
AK197832.1 | - | |||||
AK219340.1 | - | |||||
BC007467.1 | - | 3497714 | ||||
D87115.1 | - | |||||
X93150.1 | - |