Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001277932.1 | NP_001264861.1 | ||||
NM_015733.5 | NP_056548.2 | |||||
GenBank mRNA | AB019600.1 | - | ||||
AB019601.1 | - | |||||
AK085095.1 | - | |||||
AK085152.1 | - | |||||
AK085561.1 | - | |||||
AK154495.1 | - | |||||
AK156500.1 | - | |||||
AK162863.1 | - | |||||
AK203861.1 | - | |||||
BC056372.1 | - | 6834067 | ||||
BC056447.1 | - | 5721188 | ||||
BY549141.1 | - | |||||
CT010400.1 | - |