Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001130443.1 | NP_001123915.1 | ||||
NM_001130444.1 | NP_001123916.1 | |||||
NM_008284.2 | NP_032310.2 | |||||
GenBank mRNA | AJ437023.1 | - | ||||
AK183914.1 | - | |||||
AK218213.1 | - | |||||
AY373386.1 | - | |||||
BC011083.1 | - | 3152667 | ||||
BC050837.1 | - | |||||
BC061885.1 | - | |||||
BI731283.1 | - | |||||
BQ963436.1 | - | |||||
BU924724.1 | - | |||||
CK788792.1 | - |