Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ROS1 EBGEX Assays ›
Gene Symbol:
Ros1
Gene Name:
Ros1 proto-oncogene
Gene Aliases:
Ros-1, c-ros
Chromosome Location:
Chr.10: 52044865 - 52195397 on Build GRCm38
UniGene ID:
Mm.236163
Species:
Mouse
Species Specific ID (Flybase ID):
-
Type:
10
Detection Label:
Alexa Fluor 568
ProductType:
PrimeFlow Probe Set
Assay ID
VB10-3287427-PF
Size
100 tests
100 tests
Availability
Made To Order
Catalog #
PF-210
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Ros1
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_011282.2
NP_035412.2
GenBank mRNA
U15443.1
-
X81650.1
-
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
columnar/cuboidal epithelial cell development
protein phosphorylation
spermatogenesis
cell proliferation
negative regulation of gene expression
regulation of phosphate transport
cell growth
phosphorylation
peptidyl-tyrosine phosphorylation
signal transduction by protein phosphorylation
cell differentiation
regulation of TOR signaling
peptidyl-tyrosine autophosphorylation
regulation of ERK1 and ERK2 cascade
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein phosphatase binding
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0