Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_009096.3 | NP_033122.1 | ||||
GenBank mRNA | AK012284.1 | - | ||||
AK012828.1 | - | |||||
AK013333.1 | - | |||||
AK013825.1 | - | |||||
AK028205.1 | - | |||||
AK050609.1 | - | |||||
AK146952.1 | - | |||||
AK161991.1 | - | |||||
AK166680.1 | - | |||||
AK166727.1 | - | |||||
AK168050.1 | - | |||||
AK168496.1 | - | |||||
AK169624.1 | - | |||||
BC010604.1 | - | 3481640 | ||||
BC090392.1 | - | 4459055 | ||||
BC092050.1 | - | 3666815 | ||||
Y00348.1 | - |