Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_013672.2 | NP_038700.2 | ||||
GenBank mRNA | AF022363.1 | - | ||||
AF062566.1 | - | |||||
AK003347.1 | - | |||||
AK088458.1 | - | |||||
CF729889.1 | - | |||||
CJ181598.1 | - | |||||
EF200003.1 | - | |||||
EF200004.1 | - | |||||
EF200005.1 | - | |||||
EF200006.1 | - | |||||
EF200007.1 | - | |||||
EF200008.1 | - | |||||
EF200009.1 | - | |||||
S79832.1 | - |