Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001289612.1 | NP_001276541.1 | ||||
NM_001289765.1 | NP_001276694.1 | |||||
NM_001289766.1 | NP_001276695.1 | |||||
NM_009539.3 | NP_033565.2 | |||||
GenBank mRNA | AB083210.1 | - | ||||
AB084383.1 | - | |||||
AK039883.1 | - | |||||
AK089056.1 | - | |||||
BC029727.1 | - | 4925739 | ||||
BY589032.1 | - | |||||
CJ143180.1 | - | |||||
U04379.1 | - | |||||
U77667.1 | - |