Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001038663.1 | NP_001033752.1 | ||||
NM_011949.3 | NP_036079.1 | |||||
GenBank mRNA | AK013699.1 | - | ||||
AK018549.1 | - | |||||
AK035386.1 | - | |||||
AK043621.1 | - | |||||
AK048127.1 | - | |||||
AK048159.1 | - | |||||
AK087925.1 | - | |||||
AK132241.1 | - | |||||
AK148853.1 | - | |||||
AK163692.1 | - | |||||
AK193161.1 | - | |||||
AK193200.1 | - | |||||
AK205643.1 | - | |||||
AK209313.1 | - | |||||
AK210736.1 | - | |||||
AK211534.1 | - | |||||
AK216848.1 | - | |||||
AK218813.1 | - | |||||
BB648734.1 | - | |||||
BC006708.1 | - | |||||
BC058258.1 | - | 6468233 | ||||
D10939.1 | - | |||||
X58712.1 | - |