Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001142357.1 | NP_001135829.1 | ||||
NM_145477.2 | NP_663452.1 | |||||
GenBank mRNA | AJ429133.1 | - | ||||
AK053357.1 | - | |||||
AK142322.1 | - | |||||
AK179946.1 | - | |||||
AK187237.1 | - | |||||
AK209258.1 | - | |||||
BB480728.1 | - | |||||
BC021379.1 | - | 5044319 |