Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_011819.2 | NP_035949.2 | ||||
GenBank mRNA | AF159571.1 | - | ||||
AK144199.1 | - | |||||
AK159242.1 | - | |||||
AK160927.1 | - | |||||
BC067248.1 | - | 820434 |