Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1cam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008478.3 | NP_032504.3 | ||||
GenBank mRNA | AJ627046.1 | - | ||||
AK139407.1 | - | |||||
AK212283.1 | - | |||||
BC056988.1 | - | 6822217 | ||||
CJ109973.1 | - | |||||
CJ258952.1 | - | |||||
X12875.1 | - |