Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008029.3 | NP_032055.1 | ||||
GenBank mRNA | AK090148.1 | - | ||||
BC138346.1 | - | 8861366 | ||||
BC138348.1 | - | 8861368 | ||||
BU757635.1 | - | |||||
BY311090.1 | - | |||||
L07296.1 | - |