Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_017374.3 | NP_059070.1 | ||||
GenBank mRNA | AA220297.1 | - | ||||
AK090311.1 | - | |||||
BC019161.1 | - | 3674696 | ||||
BC049594.1 | - | |||||
BC058582.1 | - | 6515195 | ||||
BE847789.1 | - | |||||
CK619614.1 | - | |||||
Z67746.1 | - |