Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_011947.3 | NP_036077.1 | ||||
GenBank mRNA | AK137127.1 | - | ||||
AK142278.1 | - | |||||
AK220450.1 | - | |||||
BC023781.1 | - | 5346982 | ||||
U43187.1 | - |