Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_016896.3 | NP_058592.1 | ||||
GenBank mRNA | AF143094.1 | - | ||||
AK036200.1 | - | |||||
AK172518.1 | - | |||||
BC107015.2 | - |