Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008764.3 | NP_032790.3 | ||||
GenBank mRNA | AB013898.1 | - | ||||
AK140244.1 | - | |||||
AK146105.1 | - | |||||
BC049782.1 | - | 6506550 | ||||
U94331.1 | - |