Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001190974.1 | NP_001177903.1 | ||||
NM_001190975.1 | NP_001177904.1 | |||||
NM_009465.4 | NP_033491.2 | |||||
GenBank mRNA | AK033237.1 | - | ||||
AK048382.1 | - | |||||
AK155567.1 | - | |||||
AK161294.1 | - | |||||
AK170010.1 | - | |||||
AK170897.1 | - | |||||
AK186931.1 | - | |||||
AK199775.1 | - | |||||
AK200128.1 | - | |||||
AK200939.1 | - | |||||
BC008551.1 | - | |||||
BC046618.1 | - | 6494383 | ||||
BC050914.1 | - | 6418252 | ||||
BC058230.1 | - | 6313662 | ||||
X63535.1 | - |