Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008960.2 | NP_032986.1 | ||||
GenBank mRNA | AK009640.1 | - | ||||
AK030750.1 | - | |||||
AK037998.1 | - | |||||
AK045001.1 | - | |||||
AK051115.1 | - | |||||
AK076980.1 | - | |||||
AK080905.1 | - | |||||
AK086551.1 | - | |||||
AK087678.1 | - | |||||
AK088717.1 | - | |||||
AK138025.1 | - | |||||
AK148736.1 | - | |||||
AK164302.1 | - | |||||
AK208703.1 | - | |||||
AK212035.1 | - | |||||
AK219196.1 | - | |||||
BC021445.1 | - | 5038272 | ||||
BC150968.1 | - | 9087880 | ||||
BU701098.1 | - | |||||
CK616227.1 | - | |||||
U92437.1 | - |