Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010113.3 | NP_034243.2 | ||||
GenBank mRNA | AK084927.1 | - | ||||
AK085707.1 | - | |||||
AK088408.1 | - | |||||
AK136439.1 | - | |||||
AK142666.1 | - | |||||
AK157778.1 | - | |||||
BC017681.1 | - | 4221592 | ||||
BC060741.1 | - | 4164289 | ||||
BC092277.1 | - | 4240429 | ||||
J00380.1 | - | |||||
V00741.1 | - | |||||
X74038.1 | - |