Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001165894.1 | NP_001159366.1 | ||||
NM_009652.3 | NP_033782.1 | |||||
GenBank mRNA | AI843381.1 | - | ||||
AK040758.1 | - | |||||
AK154936.1 | - | |||||
AK196375.1 | - | |||||
AK211494.1 | - | |||||
AK214116.1 | - | |||||
AK216454.1 | - | |||||
BC066018.1 | - | 5716837 | ||||
BC115583.1 | - | |||||
KF836746.1 | - | |||||
M94335.1 | - | |||||
X65687.1 | - |