Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001163671.1 | NP_001157143.1 | ||||
NM_001163672.1 | NP_001157144.1 | |||||
NM_016961.3 | NP_058657.1 | |||||
NM_207692.2 | NP_997575.2 | |||||
GenBank mRNA | AB005664.1 | - | ||||
AF052466.1 | - | |||||
AF052467.1 | - | |||||
AF052468.1 | - | |||||
AF052469.1 | - | |||||
AK031959.1 | - | |||||
AK031974.1 | - | |||||
AK043169.1 | - | |||||
AK090057.1 | - | |||||
AK145083.1 | - | |||||
BB362351.1 | - | |||||
BC024514.1 | - | |||||
BC028341.1 | - | 4192796 | ||||
CX235052.1 | - |