Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001163215.2 | NP_001156687.1 | ||||
NM_001163216.2 | NP_001156688.1 | |||||
NM_001163217.2 | NP_001156689.1 | |||||
NM_001205270.1 | NP_001192199.1 | |||||
NM_008010.5 | NP_032036.2 | |||||
GenBank mRNA | AF024638.1 | - | ||||
AK140217.1 | - | |||||
AK143592.1 | - | |||||
AK163495.1 | - | |||||
AK163547.1 | - | |||||
BC053056.1 | - | 5708838 | ||||
BM899649.1 | - | |||||
BU053488.1 | - | |||||
BY732245.1 | - | |||||
CB247234.1 | - | |||||
CD349897.1 | - | |||||
M81342.1 | - | |||||
S56291.1 | - | |||||
X58255.1 | - |