Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008465.5 | NP_032491.2 | ||||
GenBank mRNA | AK028259.1 | - | ||||
AK028307.1 | - | |||||
AK141446.1 | - | |||||
AK154800.1 | - | |||||
AK163411.1 | - | |||||
AK163937.1 | - | |||||
AK169311.1 | - | |||||
AK169348.1 | - | |||||
AK169464.1 | - | |||||
AK195236.1 | - | |||||
AK203891.1 | - | |||||
AK218219.1 | - | |||||
BC006771.1 | - | 3588534 | ||||
U20619.1 | - | |||||
U34228.1 | - |