Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_021443.3 | NP_067418.1 | ||||
GenBank mRNA | AB023418.1 | - | ||||
AI449326.1 | - | |||||
AK007942.1 | - | |||||
BC117101.1 | - | 40126420 | ||||
BC117103.1 | - | 40126422 | ||||
BX639248.1 | - | |||||
BY707942.1 | - |