Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_080555.2 | NP_542122.1 | ||||
GenBank mRNA | AK003390.1 | - | ||||
AK011276.1 | - | |||||
AK076183.1 | - | |||||
AK159136.1 | - | |||||
AK160052.1 | - | |||||
AK160056.1 | - | |||||
AK163427.1 | - | |||||
AV018880.1 | - | |||||
BC005558.1 | - | 3964817 | ||||
BY129827.1 | - |