Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_027470.3 | NP_081746.1 | ||||
GenBank mRNA | AK017713.1 | - | ||||
AK088512.1 | - | |||||
AK129298.2 | - | |||||
AK183660.1 | - | |||||
AY217016.1 | - | |||||
BB839524.1 | - | |||||
BC030389.1 | - | |||||
BC048238.1 | - | 6475114 | ||||
BQ572351.1 | - | |||||
BY197606.1 | - | |||||
CF172418.1 | - |