Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001099276.2 | NP_001092746.2 | ||||
GenBank mRNA | AK049181.1 | - | ||||
AK165803.1 | - | |||||
BC132141.1 | - | 40130418 |