Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_017020.3 | NP_058716.2 | ||||
XM_006232587.2 | XP_006232649.1 | |||||
XM_006232588.2 | XP_006232650.1 | |||||
XM_008761127.1 | XP_008759349.1 | |||||
GenBank mRNA | CB778659.1 | - | ||||
M58587.1 | - |