Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_013005.1 | NP_037137.1 | ||||
XM_006231868.2 | XP_006231930.1 | |||||
XM_008760659.1 | XP_008758881.1 | |||||
GenBank mRNA | D63325.1 | - | ||||
D64045.1 | - | |||||
D64048.1 | - | |||||
D78486.1 | - | |||||
U50412.1 | - |