Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_138905.2 | NP_620260.2 | ||||
GenBank mRNA | BC072544.1 | - | 7105872 | |||
FQ222139.1 | - | |||||
FQ222520.1 | - | |||||
Y07783.1 | - |