Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 32 - 33 | 4945 | 67 | |
NM_180432.2 | NP_850763.1 | 31 - 32 | 4777 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |