Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105109.4 | NP_564830.1 | 4 - 5 | 679 | 75 | |
GenBank mRNA | AF370222.1 | - | 4 - 5 | 666 | 75 | |
AY059116.1 | - | 4 - 5 | 609 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |